Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MILEERPDGA GAGEESPRLQ GCDSLTQIQC GQLQSRRAQI HQQIDKELQM RTGAENLYRA TSNNRVRETV ALELSYVNSN LQLLKEELEE LSGGVDPGRH GSEAVTVPMI PLGLKETKEL DWSTPLKELI SVHFGEDGAS YEAEIRELEA LRQAMRTPSR NESGLELLTA YYNQLCFLDA RFLTPARSLG LFFHWYDSLT GVPAQQRALA FEKGSVLFNI GALHTQIGAR QDRSCTEGAR RAMEAFQRAA GAFSLLRENF SHAPSPDMSA ASLCALEQLM MAQAQECVFE GLSPPASMAP QDCLAQLRLA QEAAQVAAEY RLVHRTMAQP PVHDYVPVSW TALVHVKAEY FRSLAHYHVA MALCDGSPAT EGELPTHEQV FLQPPTSSKP RGPVLPQELE ERRQLGKAHL KRAILGQEEA LRLHALCRVL REVDLLRAVI SQTLQRSLAK YAELDREDDF CEAAEAPDIQ PKTHQKPEAR MPRLSQGKGP DIFHRLGPLS VFSAKNRWRL VGPVHLTRGE GGFGLTLRGD SPVLIAAVIP GSQAAAAGLK EGDYIVSVNG QPCRWWRHAE VVTELKAAGE AGASLQVVSL LPSSRLPSLG DRRPVLLGPR GLLRSQREHG CKTPASTWAS PRPLLNWSRK AQQGKTGGCP QPCAPVKPAP PSSLKHPGWP
Has no enzymatic activity. May serve as a target for Rho, and interact with some cytoskeletal component upon Rho binding or relay a Rho signal to other molecules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTP-Rho-binding protein 1; outer dense fiber of sperm tails 5; rhophilin 1; rhophilin, Rho GTPase binding protein 1; Rhophilin-1
Gene Aliases: KIAA1929; ODF5; RHOPHILIN; RHPN; RHPN1
UniProt ID: (Human) Q8TCX5
Entrez Gene ID: (Human) 114822
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support