Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MARSLVHDTV FYCLSVYQVK ISPTPQLGAA SSAEGHVGQG APGLMGNMNP EGGVNHENGM NRDGGMIPEG GGGNQEPRQQ PQPPPEEPAQ AAMEGPQPEN MQPRTRRTKF TLLQVEELES VFRHTQYPDV PTRRELAENL GVTEDKVRVW FKNKRARCRR HQRELMLANE LRADPDDCVY IVVD
This gene is a member of the PEPP subfamily of paired-like homoebox genes. The gene may be regulated by androgens and epigenetic mechanisms. The encoded nuclear protein is likely a transcription factor that may play a role in human reproduction. [provided by RefSeq, Dec 2012].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ovary-, testis- and epididymis-expressed gene protein; Paired-like homeobox protein PEPP-1; PEPP subfamily gene 1; Rhox homeobox family member 1; Rhox homeobox family, member 1; ZKSCAN21
Gene Aliases: OTEX; PEPP1; RHOXF1
UniProt ID: (Human) Q8NHV9
Entrez Gene ID: (Human) 158800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support