Recommended positive controls: HepG2.
Predicted reactivity: Cow: 100%; Dog: 100%; Horse: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The cascade of biochemical reactions within the vision system is complex and contains numerous steps taking place in nanoseconds. RGS is a family of GTPase accelerating proteins (GAPs). RGS9 is a member of this family and is the predominant source of GAP activity in rod outer segments. RGS9-1, found predominately in vertebrate cone and rod photoreceptors, is a useful model for understanding how rapid recovery kinetics of the light responses in retinal photoreceptors take place.