Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Aliquoting is unnecessary for -20°C storage.
The protein encoded by this gene is an RNA binding protein and possible splicing factor. The encoded protein is found in the nucleus, where it colocalizes with core spliceosomal proteins. Studies of a mouse protein with high sequence similarity to this protein suggest that this protein may act as a transcriptional coactivator for JUN/AP-1 and estrogen receptors. Multiple transcript variants encoding different isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAPER alpha; CAPERalpha; Coactivator of activating protein 1 and estrogen receptors; coactivator of activating protein-1 and estrogen receptors; Coactivator of AP-1 and ERs; functional spliceosome-associated protein 59; Hepatocellular carcinoma protein 1; RNA-binding motif protein 39; RNA-binding protein 39; RNA-binding region (RNP1, RRM) containing 2; RNA-binding region-containing 2; RNA-binding region-containing protein 2; Splicing factor HCC1; transcription coactivator CAPER
Gene Aliases: 1500012C14Rik; 2310040E03Rik; B330012G18Rik; C79248; CAPER; CAPERalpha; FSAP59; HCC1; R75070; RBM39; Rnp1; RNPC2; Rrm
UniProt ID: (Human) Q14498, (Mouse) Q8VH51
Entrez Gene ID: (Human) 9584, (Rat) 362251, (Mouse) 170791
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support