Predicted to react with bovine, chimpanzee, dog, rabbit and rhesus monkey based on sequence homology.
Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for immunohistochemical applications is diabetic kidney. In Western blot, this antibody detects a band at ~45 kDa in mouse and rat lung lysates.
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.