Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MEAGGFLDSL IYGACVVFTL GMFSAGLSDL RHMRMTRSVD NVQFLPFLTT EVNNLGWLSY GALKGDGILI VVNTVGAALQ TLYILAYLHY CPRKAKVIQT KSTQCLSYPL TIATLLTSAS WCLYGFRLRD PYIMVSNFPG IVTSFIRFWL FWKYPQEQDR NYWLLQT
SLC50A1 gene ontology annotations related to this gene include carbohydrate transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsSWEET1; probable sugar transporter RAG1AP1; RAG1-activating protein 1; recombination activating gene 1 activating protein 1; RP11-540D14.5; RZPDo834D038D; solute carrier family 50 (sugar efflux transporter), member 1; solute carrier family 50 (sugar transporter), member 1; Solute carrier family 50 member 1; Stromal cell protein; Sugar transporter SWEET1
Gene Aliases: HsSWEET1; RAG1AP1; SCP; SLC50A1; slv; SWEET1
UniProt ID: (Human) Q9BRV3
Entrez Gene ID: (Human) 55974
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support