Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEVRASLQKI VSNGDEQLEK AMEEILRDSE KRPSSLLVDC QSSSEISDHS FGDIPASQTN KPSLQLILDP SNTEISTPRP SSPGGLPEED SVLFNKLTYL GCMKVSSPRN EVEALRAMAT MKSSSQYPFP VTLYVPNVPE GSVRIIDQSS NVEIASFPIY KVLFCARGHD GTTESNCFAF TESSHGSEEF QIHVFSCEIK EAVSRILYSF CTAFKRSSRQ VSDVKDSVIP TPDSDVFTFS VSLEVKEDDG KGNFSPVPKD RDKFYFKLKQ GIEKKVVITV QQLSNKELAI ERCFGMLLSP GRNVKNSDMH LLDMESMGKS YDGRAYVITG MWNPNAPVFL ALNEETPKDK QVYMT
RABGAP1L is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: expressed in hematopoietic cells, heart, liver (HLL); RAB GTPase activating protein 1-like; Rab GTPase-activating protein 1-like; Rab GTPase-activating protein 1-like, isoform 10; RABGAP1L; TBC1 domain family, member 18
Gene Aliases: HHL; KIAA0471; RABGAP1L; TBC1D18
UniProt ID: (Human) B7ZAP0
Entrez Gene ID: (Human) 9910
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support