Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Pre-Pro-Vasopressin Antibodies

          Invitrogen

          Pre-Pro-Vasopressin Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Pre-Pro-Vasopressin Polyclonal Antibody

          Product Details

          PA5-143640

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to key amino acid residues surrounding the cleavage sites for the human vasopressin precursor peptide, conjugated to keyhole limpet hemocyanin (KLH).

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          10mM HEPES, pH 7.5, with 100µg/mL BSA, 50% glycerol, 150mM NaCl

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942868

          Product Specific Information

          Specific for locally produced levels of the ~ 20 kDa uncleaved pre-pro-vasopressin protein. Immunolabeling of the uncleaved pre-pro-vasopressin protein band is completely blocked by preadsorption of the antibody with the peptide used as antigen.

          Target Information

          Vasopressin is a nine amino-acid peptide hormone that plays a key role in water and blood pressure homeostasis (Qureshi S., et al 2014). Vasopressin is the end-product of a highly processed 164 amino acid pre-pro-peptide. Processing of the vasopressin pre-pro-peptide results in three distinct peptides with a 1:1:1 ratio: vasopressin, neurophysin II, and copeptin (Arroyo J.P. et al, 2021). Vasopressin is the biologically active hormone, neurophysin II is a carrier molecule for vasopressin, and copeptin is the c-terminal glycosylated end-product. Vasopressin has been thought to be primarily made in the brain, and the sole source of vasopressin stimulating vasopressin V2 receptors in the kidney until recent studies (Arroyo JP, et al 2022). This antibody specially detects uncleaved pre-pro-vasopressin, accurately identifying locally produced vasopressin versus peripheral uptake of hypothalamic vasopressin or related peptides (Arroyo J.P. et al, 2022). This key research tool aided the Arroyo group in discovering immunoreactive vasopressin outside of the brain by recognizing localized vasopressin in human and mouse kidney, specifically in the distal nephron.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: antidiuretic hormone; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); ARVP; AVP-NPII; copeptin; neurohypophyseal; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide; VP

          View more View less

          Gene Aliases: ADH; ARVP; AVP; AVP-NPII; AVRP; DI; Vas; VP; Vsp

          View more View less

          UniProt ID: (Human) P01185, (Mouse) P35455, (Rat) P01186

          View more View less

          Entrez Gene ID: (Human) 551, (Mouse) 11998, (Rat) 24221

          View more View less

          Function(s)
          protein kinase activity signal transducer activity receptor binding neuropeptide hormone activity neurohypophyseal hormone activity V1A vasopressin receptor binding V1B vasopressin receptor binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process hormone activity protein binding V2 vasopressin receptor binding
          Process(es)
          maternal aggressive behavior positive regulation of systemic arterial blood pressure signal transduction positive regulation of cytosolic calcium ion concentration negative regulation of female receptivity grooming behavior locomotory behavior positive regulation of cell proliferation positive regulation of gene expression positive regulation of glutamate secretion positive regulation of cell growth positive regulation of cAMP biosynthetic process positive regulation of prostaglandin biosynthetic process positive regulation of cellular pH reduction positive regulation of peptidyl-serine phosphorylation social behavior regulation of renal sodium excretion vasoconstriction maternal behavior negative regulation of apoptotic process penile erection negative regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of vasoconstriction regulation of blood vessel size multicellular organismal water homeostasis negative regulation of transmission of nerve impulse ERK1 and ERK2 cascade protein kinase C signaling negative regulation of release of cytochrome c from mitochondria renal water homeostasis generation of precursor metabolites and energy water transport apoptotic process cell-cell signaling response to testosterone response to nicotine hyperosmotic salinity response sodium-independent organic anion transport response to ethanol temperature homeostasis protein phosphorylation smooth muscle contraction response to organic cyclic compound
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0