Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PTP1B Antibodies

          Invitrogen

          Phospho-PTP1B (Tyr152) Polyclonal Antibody

          View all (49) PTP1B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-PTP1B (Tyr152) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-PTP1B (Tyr152) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PTP1B (Tyr152) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PTP1B (Tyr152) Antibody (PA5-143755) in WB

          Western blot of Phospho-PTP1B (Tyr152) in mouse SYF cSrc-transformed cells untreated (lanes 1 & 3) or treated (lanes 2 & 4) with pervanadate (1 mM for 30 minutes). The blots were probed with rabbit polyclonal anti-PTP1B (a.a. 146-157) (lanes 1 & 2) or Phospho-PTP1B (Tyr152) polyclonal antibody (Product # PA5-143755) (lanes 3 & 4). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PTP1B (Tyr152) Antibody in Western Blot (WB)

          Product Details

          PA5-143755

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PTP1B (Tyr-152) synthetic peptide corresponds to amino acids surrounding Tyr-152 in human PTP1B. This sequence has significant homology to the conserved site in rat and mouse PTP1B, and has homology to conserved sites in TC-PTP and MPTP, but not other protein tyrosine phosphatase family members.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942983

          Product Specific Information

          This antibody was cross-adsorbed to a phospho-tyrosine peptide then affinity purified with phospho-PTP1B (Tyr-152) peptide. The purified antibody detects a 50 kDa* band corresponding to PTP1B in western blots of pervanadate treated human A431 and Jurkat, as well as mouse SYF cells. This reactivity is not observed in control cells or after alkaline phosphatase treatment of blots.

          Target Information

          PTP1B (PTPN1) is a tyrosine-protein phosphatase which acts as a regulator of endoplasmic reticulum unfolded protein response. It mediates dephosphorylation of EIF2AK3/PERK and in turn inactivates the protein kinase activity of EIF2AK3/PERK. PTP1B also plays a role in CKII- and p60c-src-induced signal transduction cascades. It regulates the EFNA5-EPHA3 signaling pathway which modulates cell reorganization and cell-cell repulsion. PTP1B also regulates the hepatocyte growth factor receptor signaling pathway through dephosphorylation of MET.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CPTP1; EC 3.1.3.48; hgnc9642; KIAA0283; protein tyrosine phosphatase, placental; protein-protein-tyrosine phosphatase HA2; Protein-tyrosine phosphatase 1B; Protein-tyrosine phosphatase HA2; PTN1; PTP-1B; PTP-HA2; RP5-1121H13.2; RPTPrho; tyrosine phosphatase 1b; tyrosine-phosphatase; Tyrosine-protein phosphatase non-receptor type 1

          View more View less

          Gene Aliases: Ptp; PTP-1B; PTP-HA2; PTP1B; PTPN1

          View more View less

          UniProt ID: (Human) P18031, (Rat) P20417, (Mouse) P35821

          View more View less

          Entrez Gene ID: (Human) 5770, (Rat) 24697, (Mouse) 19246

          View more View less

          Function(s)
          protein tyrosine phosphatase activity insulin receptor binding protein binding zinc ion binding enzyme binding protein kinase binding receptor tyrosine kinase binding poly(A) RNA binding ephrin receptor binding protein phosphatase 2A binding cadherin binding involved in cell-cell adhesion phosphoprotein phosphatase activity hydrolase activity phosphatase activity
          Process(es)
          protein dephosphorylation insulin receptor signaling pathway regulation of signal transduction regulation of endocytosis negative regulation of vascular endothelial growth factor receptor signaling pathway endoplasmic reticulum unfolded protein response actin cytoskeleton reorganization peptidyl-tyrosine dephosphorylation platelet-derived growth factor receptor-beta signaling pathway regulation of insulin receptor signaling pathway negative regulation of ERK1 and ERK2 cascade regulation of hepatocyte growth factor receptor signaling pathway peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity activation of signaling protein activity involved in unfolded protein response activation of JUN kinase activity negative regulation of signal transduction regulation of intracellular protein transport IRE1-mediated unfolded protein response negative regulation of MAP kinase activity negative regulation of insulin receptor signaling pathway regulation of type I interferon-mediated signaling pathway JAK-STAT cascade involved in growth hormone signaling pathway positive regulation of protein tyrosine kinase activity cell-cell adhesion negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of IRE1-mediated unfolded protein response negative regulation of PERK-mediated unfolded protein response positive regulation of receptor catabolic process dephosphorylation response to endoplasmic reticulum stress
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fd7d668bc-tpg7z:80/100.66.134.75:80.
          git-commit: 4d16a52b65a0a59161c0e81e148ee2d2ac6d3307
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc