PA1-26733 detects PKA beta (catalytic subunit) (phospho S338) from human and mouse samples. PA1-26733 is expected to cross react with porcine and bovine due to sequence homology.
PA1-26733 has been successfully used in Western blot procedures.
The PA1-26733 immunogen is a synthetic phosphopeptide corresponding to a portion of human PKA catalytic beta subunit containing serine 338.
The catalytic subunit C-beta of PKA (PRKACB) is a member of the Ser/Thr protein kinase family (the PKA catalytic subunit consist of three gene products: C-alpha, C-beta, and C-gamma) and has been assigned to human chromosome region 1p36.1. PRKACB is derived from a gene distinct from C-alpha and shows tissue-specific expression. At the amino acid level C-alpha and C-beta showed 93% homology. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans.