Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NMDAR1 Antibodies

          Invitrogen

          Phospho-NMDAR1 (Ser896, Ser897) Polyclonal Antibody

          View all (45) NMDAR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-NMDAR1 (Ser896, Ser897) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-NMDAR1 (Ser896, Ser897) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-NMDAR1 (Ser896, Ser897) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-NMDAR1 (Ser896, Ser897) Antibody (PA5-121322) in WB

          Western blot analysis of Phospho-NMDAR1 (Ser896, Ser897) in extracts of HeLa cells. Samples were incubated with Phospho-NMDAR1 (Ser896, Ser897) Polyclonal antibody (Product # PA5-121322) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. HeLa cells were treated by UV at room temperature for 30 minutes and PMA (200 nM) at 37°C for 30 minutes ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-NMDAR1 (Ser896, Ser897) Antibody in Western Blot (WB)

          Product Details

          PA5-121322

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic phosphorylated peptide around S896 & S897 of human NMDAR1 (NP_015566.1).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.67 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2914894

          Product Specific Information

          Positive test controls include: HeLa. The target is usually found in the following locations: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, postsynaptic density, synapse.

          Immunogen sequence: RRSSKD

          Target Information

          NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: GluN1; Glun1r; glutamate [NMDA] receptor subunit zeta 1; Glutamate [NMDA] receptor subunit zeta-1; glutamate receptor; Glutamate receptor ionotropic, NMDA 1; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDZ1; NMZ1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; RP11-350O14.1

          View more View less

          Gene Aliases: GluN1; GluRdelta1; Glurz1; GluRzeta1; GRIN1; M100174; MRD8; NMD-R1; NMDA1; Nmdar; NMDAR1; NR1; Rgsc174

          View more View less

          UniProt ID: (Human) Q05586, (Rat) P35439, (Mouse) P35438

          View more View less

          Entrez Gene ID: (Human) 2902, (Rat) 24408, (Mouse) 14810

          View more View less

          Function(s)
          NMDA glutamate receptor activity Ras guanyl-nucleotide exchange factor activity extracellular-glutamate-gated ion channel activity calcium channel activity calcium ion binding protein binding calmodulin binding glycine binding glutamate binding enzyme binding voltage-gated cation channel activity glutamate receptor binding neurotransmitter binding protein heterodimerization activity ionotropic glutamate receptor activity receptor binding cation channel activity protein dimerization activity receptor activity ion channel activity
          Process(es)
          MAPK cascade conditioned taste aversion suckling behavior response to amphetamine cation transport cellular calcium ion homeostasis chemical synaptic transmission respiratory gaseous exchange long-term memory adult locomotory behavior olfactory learning visual learning propylene metabolic process sensory perception of pain pons maturation cerebral cortex development regulation of ion transmembrane transport social behavior ionotropic glutamate receptor signaling pathway synaptic transmission, glutamatergic regulation of membrane potential positive regulation of apoptotic process response to morphine negative regulation of neuron apoptotic process positive regulation of GTPase activity regulation of respiratory gaseous exchange response to ethanol positive regulation of transcription from RNA polymerase II promoter ephrin receptor signaling pathway regulation of long-term neuronal synaptic plasticity rhythmic process regulation of dendrite morphogenesis regulation of axonogenesis protein tetramerization response to calcium ion regulation of synapse assembly calcium ion homeostasis excitatory postsynaptic potential prepulse inhibition male mating behavior response to fungicide calcium ion transmembrane transport cellular response to manganese ion positive regulation of excitatory postsynaptic potential startle response calcium ion transport learning or memory learning memory associative learning regulation of cell communication positive regulation of cell death response to organic cyclic compound response to amine ion transmembrane transport regulation of neuron apoptotic process regulation of synaptic plasticity regulation of neuronal synaptic plasticity neuromuscular process transport ion transport cation transmembrane transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fd7d668bc-tpg7z:80/100.66.134.75:80.
          git-commit: 4d16a52b65a0a59161c0e81e148ee2d2ac6d3307
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc