Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CDK5 Antibodies

          Bioss

          Phospho-CDK5 (Tyr15) Polyclonal Antibody

          View all (32) CDK5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-CDK5 (Tyr15) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-CDK5 (Tyr15) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-CDK5 (Tyr15) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-CDK5 (Tyr15) Antibody (BS-6411R) in IHC (P)

          Formalin-fixed and paraffin embedded mouse embryo tissue labeled with Rabbit Anti phospho-CDK5(Tyr15) Polyclonal Antibody, Unconjugated (bs-6411R) at 1:200 followed by conjugation to the secondary antibody and DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-CDK5 (Tyr15) Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          BS-6411R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic phosphopeptide derived from human CDK5 around the phosphorylation site of Tyr15.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          CDK5 is a member of the Cyclin-Dependent Kinase family that is most abundant in the mammalian brain. Active form of CDK5, which has also been called neuronal cdc2-like kinase, is a heterodimer of CDK5 and a 25 kDa protein which is derived proteolytically from a 35 kDa brain and neuron-specific protein and is essential for the kinase activity of CDK5. CDK5 has emerged as a crucial regulator of neuronal migration in the developing central nervous system. CDK5 phosphorylates a diverse list of substrates, implicating it in the regulation of a range of cellular processes - from adhesion and motility, to synaptic plasticity and drug addiction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CDK5 activator 1; CDK5-SV; Cell division protein kinase 5; CR6 protein kinase; Cyclin-dependent kinase 5; Cyclin-dependent-like kinase 5; protein kinase CDK5 splicing; Serine/threonine-protein kinase PSSALRE; Tau protein kinase II catalytic subunit; TPKII catalytic subunit

          View more View less

          Gene Aliases: AW048668; CDK5; CDKN5; Crk6; LIS7; PSSALRE

          View more View less

          UniProt ID: (Human) Q00535, (Mouse) P49615, (Rat) Q03114

          View more View less

          Entrez Gene ID: (Human) 1020, (Mouse) 12568, (Rat) 140908

          View more View less

          Function(s)
          p53 binding protein kinase activity protein serine/threonine kinase activity cyclin-dependent protein serine/threonine kinase activity ErbB-2 class receptor binding protein binding ATP binding cytoskeletal protein binding kinase activity protein kinase binding acetylcholine receptor activator activity ErbB-3 class receptor binding ephrin receptor binding tau-protein kinase activity nucleotide binding transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          neuron migration synaptic transmission, dopaminergic intracellular protein transport nucleocytoplasmic transport cell cycle cell-matrix adhesion chemical synaptic transmission synapse assembly skeletal muscle tissue development motor neuron axon guidance cell proliferation visual learning response to wounding Schwann cell development synaptic vesicle exocytosis regulation of macroautophagy phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation sensory perception of pain cerebellar cortex formation hippocampus development layer formation in cerebral cortex central nervous system neuron development corpus callosum development neuron differentiation regulation of cell migration negative regulation of axon extension cortical actin cytoskeleton organization neuron projection development negative regulation of protein ubiquitination negative regulation of synaptic plasticity positive regulation of protein binding receptor catabolic process serine phosphorylation of STAT3 protein synaptic transmission, glutamatergic protein localization to synapse regulation of apoptotic process receptor clustering positive regulation of neuron apoptotic process negative regulation of cell cycle positive regulation of protein kinase activity negative regulation of proteolysis negative regulation of transcription, DNA-templated positive regulation of calcium ion-dependent exocytosis protein autophosphorylation negative regulation of protein export from nucleus behavioral response to cocaine regulation of synaptic plasticity synaptic vesicle endocytosis rhythmic process axon extension oligodendrocyte differentiation dendrite morphogenesis cell division neuron apoptotic process excitatory postsynaptic potential regulation of dendritic spine morphogenesis calcium ion import regulation of cell cycle arrest positive regulation of protein targeting to membrane negative regulation of neuron death regulation of signal transduction by p53 class mediator regulation of synaptic vesicle recycling positive regulation of actin cytoskeleton reorganization positive regulation of receptor activity protein phosphorylation exocytosis apoptotic process nervous system development axonogenesis associative learning cell migration telencephalon development cerebellum development cerebellar cortex development cerebral cortex development forebrain development response to cocaine regulated exocytosis neuron projection morphogenesis regulation of cell cycle regulation of postsynaptic membrane potential central nervous system development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5fd7d668bc-tpg7z:80/100.66.134.75:80.
          git-commit: 4d16a52b65a0a59161c0e81e148ee2d2ac6d3307
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc