Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CAMKK alpha/beta Antibodies

          Invitrogen

          Phospho-CAMKK alpha/beta (Ser458, Ser495) Polyclonal Antibody

          View all (2) CAMKK alpha/beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-CAMKK alpha/beta (Ser458, Ser495) Polyclonal Antibody

          • Antibody Testing Data (10)
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody (PA5-105869) in IHC (P)

          Immunohistochemistry analysis of Phospho-CAMKK alpha/beta (Ser458, Ser495) in human gastric cancer and adjacent normal tissues. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with Phospho-CAMKK alpha/beta (Ser458, Ser495) polyclonal antibody (Product # PA5-105869) using a dilution of 1:100 (4°C overnight) followed by HRP c... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Western Blot (WB)
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Western Blot (WB)
          Phospho-CAMKK alpha/beta (Ser458, Ser495) Antibody in Western Blot (WB)

          Product Details

          PA5-105869

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CAMKK1(Accession Q8N5S9), corresponding to amino acid residues around phosphorylated Ser458.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2817268

          Product Specific Information

          Antibody detects endogenous levels of CAMKK1/2 only when phosphorylated at Ser458/495.

          Target Information

          The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This protein plays a role in the calcium/calmodulin-dependent (CaM) kinase cascade. Three transcript variants encoding two distinct isoforms have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha CaMKK; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium calmodulin dependent protein kinase kinase alpha; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; Calcium/calmodulin-dependent protein kinase kinase 1; calcium/calmodulin-dependent protein kinase kinase 1, alpha; Calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; Calcium/calmodulin-dependent protein kinase kinase alpha; Calcium/calmodulin-dependent protein kinase kinase beta; CaM-kinase IV kinase; caM-kinase kinase 1; caM-kinase kinase 2; caM-kinase kinase alpha; CaM-kinase kinase beta; CaM-KK 1; CaM-KK 2; CaM-KK alpha; CaM-KK beta; caMKK 1; caMKK 2; caMKK alpha; CAMKK alpha protein; caMKK beta; CAMKK beta protein

          View more View less

          Gene Aliases: 6330570N16Rik; AI846603; AW061083; CAMKK; CAMKK1; CAMKK2; CAMKKA; CaMKKalpha; CAMKKB; KIAA0787; mKIAA0787

          View more View less

          UniProt ID: (Human) Q8N5S9, (Human) Q96RR4, (Mouse) Q8VBY2, (Rat) P97756, (Rat) O88831, (Mouse) Q8C078

          View more View less

          Entrez Gene ID: (Human) 84254, (Human) 10645, (Mouse) 55984, (Rat) 60341, (Rat) 83506, (Mouse) 207565

          View more View less

          Function(s)
          calmodulin-dependent protein kinase activity protein binding calmodulin binding ATP binding protein tyrosine kinase activity calcium ion binding nucleotide binding protein kinase activity protein serine/threonine kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups protein kinase activator activity
          Process(es)
          MAPK cascade protein phosphorylation peptidyl-tyrosine phosphorylation calcium-mediated signaling regulation of protein kinase activity positive regulation of transcription, DNA-templated protein autophosphorylation activation of protein kinase activity phosphorylation positive regulation of protein kinase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0