Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Acetyl-CoA Carboxylase Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          Phospho-Acetyl-CoA Carboxylase (Ser80) Polyclonal Antibody

          View all (39) Acetyl-CoA Carboxylase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Datasheet
          Protocols

          Cite Phospho-Acetyl-CoA Carboxylase (Ser80) Polyclonal Antibody

          • Antibody Testing Data (23)
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 23

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody (PA5-104981) in ICC/IF

          Immunocytochemistry analysis of Phospho-Acetyl-CoA Carboxylase (Ser80) in H2O2 treated A549 cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with Phospho-Acetyl-CoA Carboxylase (Ser80) polyclonal antibody (Product # PA5-104981) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conju... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Acetyl-CoA Carboxylase (Ser80) Antibody in Western Blot (WB)

          Product Details

          PA5-104981

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human ACACA(Accession Q13085), corresponding to amino acid residues around phosphorylated Ser80.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816454

          Product Specific Information

          Antibody detects endogenous levels of ACC1 only when phosphorylated at Ser80, which site historically referenced as Ser79.

          Target Information

          Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0