Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: TAGTPEVWVQ VRMEATELSS FTIRCGFLGS GSISLVTVSW GGPDGAGGTT LAVLHPERGI RQWAPARQAR WETQSSISLI LEGSGASSPC ANTTFCCKFA SFPEGSWEAC GSLPPSSDPG LSAPPTPAPI LRADLAGILG VSGVLLFGCV YLLHLLRRHK HRPAPRLQPS RTSPQAPRAR AWAPSQASQA ALHVPYATIN TSCRPATLDT AHPHGGPSWW APLPTHAAHR PQGPAAWAST PIPARGSFVS VENGLYAQAG ERPPHTGPGL TLFPDPRGPR AMEGPLGVR
Cell surface receptor for NECTIN2. May act as a coinhibitory receptor that suppresses T-cell receptor-mediated signals. Following interaction with NECTIN2, inhibits T-cell proliferation. Competes with CD226 for NECTIN2-binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD112 receptor; CD112R; Poliovirus receptor-related immunoglobulin domain-containing protein; Transmembrane protein PVRIG
Gene Aliases: C7orf15; CD112R; PVRIG
UniProt ID: (Human) Q6DKI7
Entrez Gene ID: (Human) 79037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support