Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PTPRN Antibodies

          Abnova

          PTPRN Polyclonal Antibody, MaxPab™

          View all (13) PTPRN antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PTPRN Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          PTPRN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PTPRN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTPRN Antibody (H00005798-B02P) in WB

          Western Blot analysis of PTPRN expression in transfected 293T cell line by PTPRN MaxPab polyclonal antibody. Lane 1: PTPRN transfected lysate(104.5 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTPRN Antibody in Western Blot (WB)

          Product Details

          H00005798-B02P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PTPRN (AAH70053.1, 1 a.a. approximately 950 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MRRPRRPGGL GGSGGLRLLL CLLLLSSRPG GCSAVSAHGC LFDRRLCSHL EVCIQDGLFG QCQVGVGQAR PLLQVTSPVL QRLQGVLRQL MSQGLSWHDD LTQYVISQEM ERIPRLRPPE PRPRDRSGLA PKRPGPAGEL LLQDIPTGSA PAAQHRLPQP PVGKGGAGAS SSLSPLQAEL LPPLLEHLLL PPQPPHPSLS YEPALLQPYL FHQFGSRDGS RVSEGSPGMV SVGPLPKAEA PALFSRTASK GIFGDHPGHS YGDLPGPSPA QLFQDSGLLY LAQELPAPSR ARVPRLPEQG SSSRAEDSPE GYEKEGLGDR GEKPASPAVQ PDAALQRLAA VLAGYGVELR QLTPEQLSTL LTLLQLLPKG AGRNPGGVVN VGADIKKTME GPVEGRDTAE LPARTSPMPG HPTASPTSSE VQQVPSPVSS EPPKAARPPV TPVLLEKKSP LGQSQPTVAG QPSARPAAEE YGYIVTDQNV VGPALTFRIR HNEQNLSLAD VTQQAGLVKS ELEAQTGLQI LQTGVGQREE AAAVLPQTAH STSPMRSVLL TLVALAGVAG LLVALAVALC VRQHARQQDK ERLAALGPEG AHGDTTFEYQ DLCRQHMATK SLFNRAEGPP EPSRVSSVSS QFSDAAQASP SSHSSTPSWC EEPAQANMDI STGHMILAYM EDHLRNRDRL AKEWQALCAY QAEPNTCATA QGEGNIKKNR HPDFLPYDHA RIKLKVESSP SRSDYINASP IIEHDPRMPA YIATQGPLSH TIADFWQMVW ESGCTVIVML TPLVEDGVKQ CDRYWPDEGA SLYHVYEVNL VSEHIWCEDF LVRSFYLKNV QTQETRTLTQ FHFLSWPAEG TPASTRPLLD FRRKVNKCYR GRSCPIIVHC SDGAGRTGTY ILIDMVLNRM AKGVKEIDIA ATLEHVRDQR PGLVRSKDQF EFALTAVAEE VNAILKALPQ

          Target Information

          IA-2 (insulinoma-associated protein 2, ICA512, PTPRN) is a member of the transmembrane protein tyrosine phosphatase (PTP) family located in secretory granules of neuroendocrine cells. The IA-2 protein is 979 amino acids in length and consists of a luminal domain, transmembrane domain, and cytoplasmic domain. Although a member of the PTP family, IA-2 lacks phosphatase activity with known substrates due to amino acid substitutions at critical sites in its PTP domain. Two paralog RPTPs, IA-2 and IA-2beta were identified as major autoantigens in type-1 diabetes mellitus. On granule exocytosis, the IA-2 cytoplasmic domain is cleaved and the resulting cytosolic fragment moves into the nucleus where it enhances the levels of phosphorylated STAT5 and STAT3, thereby inducing insulin gene transcription and granule biogenesis. IA-2 signaling enhances pancreatic beta-cell proliferation by regulating cyclins D through STATs.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ICA 512; islet cell antigen 2; Islet cell antigen 512; Islet cell autoantigen 3; protein tyrosine phosphatase, receptor type, N; protein tyrosine phosphatase-like N; PTP IA-2; R-PTP-N; Receptor-type tyrosine-protein phosphatase-like N

          View more View less

          Gene Aliases: IA-2; IA-2/PTP; IA2; ICA3; ICA512; PTPRN; R-PTP-N

          View more View less

          UniProt ID: (Human) Q16849

          View more View less

          Entrez Gene ID: (Human) 5798

          View more View less

          Function(s)
          protein tyrosine phosphatase activity protein binding spectrin binding GTPase binding
          Process(es)
          response to reactive oxygen species luteinization cytokine-mediated signaling pathway insulin secretion response to insulin insulin secretion involved in cellular response to glucose stimulus response to estrogen response to cAMP dense core granule maturation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7dfb8678b5-vqhv7:80/100.66.129.46:80.
          git-commit: 632cddff25488e9c1007a93c87c1899f725eef0d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0