Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MSRSLDSARS FLERLEARGG REGAVLAGEF SKRCERYWAQ EQEPLQTGLF CITLIKEKWL NEDIMLRTLK VTFQKESRSV YQLQYMSWPD RGVPSSPDHM LAMVEEARRL QGSGPEPLCV HCSAGCGRTG VLCTVDYVRQ LLLTQMIPPD FSLFDVVLKM RKQRPAAVQT EEQYRFLYHT VAQMFCSTLQ NASPHYQNIK ENCAPLYDDA; Positive Samples: U-251MG, SW620; Cellular Location: Cytoplasm, Nucleus
Differentially dephosphorylate autophosphorylated tyrosine kinases which are known to be overexpressed in tumor tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-derived phosphatase; protein tyrosine phosphatase, non-receptor type 18 (brain-derived); Tyrosine-protein phosphatase non-receptor type 18
Gene Aliases: BDP1; PTP-HSCF; PTPN18
UniProt ID: (Human) Q99952
Entrez Gene ID: (Human) 26469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support