Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEQVEILRKF IQRVQAMKSP DHNGEDNFAR DFMRLRRLST KYRTEKIYPT ATGEKEENVK KNRYKDILPF DHSRVKLTLK TPSQDSDYIN ANFIKGVYGP KAYVATQGPL ANTVIDFWRM IWEYNVVIIV MACREFEMGR KKCERYWPLY GEDPITFAPF KISCEDEQAR TDYFIRTLLL EFQNESRRLY QFHYVNWPDH DVPSSFDSIL DMISLMRKYQ EHEDVPICIH CSAGCGRTGA ICAIDYTWNL LKAGKIPEEF NVFNLIQEMR TQRHSAVQTK EQYELVHRAI AQLFEKQLQL YEIHGAQKIA DGVNEINTEN MISSIEPEKQ DSPPPKPPRT RSCLVEGDAK EEILQPPEPH PVPPILTPSP PSAFPTVTTV WQDNDRYHPK PVLHMVSSEQ HSADLNRNYS KSTELPGKNE STIEQIDKKL ERNLSFEIKK VPLQEGPKSF DGNTLLNRGH AIKIKSASPC IADKISKPQE LSSDLNVGDT SQNSCVDCSV TQSNKVSVTP PEESQNSDTP PRPDRLPLDE KGHVTWSFHG PENAIPIPDL SEGNSSDINY QTRKTVSLTP SPTTQVETPD LVDHDNTSPL FRTPLSFTNP LHSDDSDSDE RNSDGAVTQN KTNISTASAT VSAATSTESI STRKVLPMSI ARHNIAGTTH SGAEKDVDVS EDSPPPLPER TPESFVLASE HNTPVRSEWS ELQSQERSEQ KKSEGLITSE NEKCDHPAGG IHYEMCIECP PTFSDKREQI SENPTEATDI GFGNRCGKPK GPRDPPSEWT
PTP PEST is an 88 kDa cytosolic protein tyrosine phosphatase which is ubiquitiously expressed in mammalian tissues. PTP PEST exhibits a high specific activity when assayed in vitro with artificial tyrosine-phosphorylated substrates. PTP PEST is subject to regulation via phosphorylation of ser 39 and is catalyzed by both protein kinase C and protein kinase A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: pest; Protein-tyrosine phosphatase G1; PTP-PEST; PTPG1; tcag7.1075; Tyrosine-protein phosphatase non-receptor type 12
Gene Aliases: PTP-PEST; PTPG1; PTPN12
UniProt ID: (Human) Q05209
Entrez Gene ID: (Human) 5782
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support