Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PTGES2 Antibodies

          Invitrogen

          PTGES2 Monoclonal Antibody (K06332_11H12)

          View all (18) PTGES2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PTGES2 Monoclonal Antibody (K06332_11H12)

          • Antibody Testing Data (2)
          PTGES2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PTGES2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTGES2 Antibody (MA5-55837) in WB

          Western blot analysis of PTGES2 in 15 µg of HeLa. Sample was run on 6-18% SDS-PAGE under reducing conditions, blotted onto nitrocellulose membrane, and peroxidase conjugated goat anti-mouse IgG was used as the secondary antibody. PTGES2 band was visualized using ECL Western Blotting Substrate. Incubation with primary PTGES2 monoclonal antibody (Product # MA5-55837) at a dilution of 1 µg/mL was used. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTGES2 Antibody in Western Blot (WB)
          PTGES2 Antibody in Immunoprecipitation (IP)

          Product Details

          MA5-55837

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunoprecipitation (IP)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          K06332_11H12

          Immunogen

          Recombinant human PTGES2 (immunogen range is 91-370).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          RRID

          AB_3668069

          Product Specific Information

          Sequence of this protein is as follows: AAQLSLSSRL QLTLYQYKTC PFCSKVRAFL DFHALPYQVV EVNPVRRAEI KFSSYRKVPI LVAQEGESSQ QLNDSSVIIS ALKTYLVSGQ PLEEIITYYP AMKAVNEQGK EVTEFGNKYW LMLNEKEAQQ VYGGKEARTE EMKWRQWADD WLVHLISPNV YRTPTEALAS FDYIVREGKF GAVEGAVAKY MGAAAMYLIS KRLKSRHRLQ DNVREDLYEA ADKWVAAVGK DRPFMGGQKP NLADLAVYGV LRVMEGLDAF DDLMQHTHIQ PWYLRVERAI

          Target Information

          Prostoglandin E2 (PGE2) is a potent lipid mediator produced by metabolism of arachidonic acid via the cyclooxygenase (COX) pathway. Following cellular stimulation, arachidonic acid is hydrolyzed from phospholipids stores by one of a family of phospholipase A2 enzymes and converted to the unstable endoperoxide PGH2 by either COX 1 or COX 2. PGH2 is isomerized to PGE2 by one of 3 distinct enzymes microsomal PGES 1 (mPGES 1), mPGES 2, or a cytosolic enzyme, cPGES. PGE2 is active in a number of physiological systems and settings including inflammation, immune regulation, generation of fever, pain perception, protection of the gastric muscosa, fertility and parturition, as well as sodium and water retention. The effects of PGE2 are transduced by 4 subtypes of G protein coupled receptors designated EP1, EP2, EP3, EP4, with Kd values for PGE2 ranging from 1 10 nM. PGE2 is rapidly metabolized in vivo by the prostaglandin 15 dehydrogenase pathway (15 hydroxy PGDH) to the inactive metabolite 13,14 dihydro 15 keto PGE2. The half life of PGE2 in the circulatory system is ~30 seconds and normal plasma levels are 3 12 pg/mL

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ14038; gamma-interferon-activated transcriptional element-binding factor 1; GATE-binding factor 1; membrane-associated prostaglandin E synthase 2; Membrane-associated prostaglandin E synthase-2; MGC11289; Microsomal prostaglandin E synthase 2; microsomal prostaglandin E synthase-2; mPGE synthase-2; mPGES-2; Prostaglandin E synthase 2; Prostaglandin-H(2) E-isomerase; RP11-395P17.2

          View more View less

          Gene Aliases: C9orf15; GBF-1; GBF1; mPGES-2; PGES2; PTGES2

          View more View less

          UniProt ID: (Human) Q9H7Z7

          View more View less

          Entrez Gene ID: (Human) 80142

          View more View less

          Function(s)
          DNA binding protein binding electron carrier activity protein disulfide oxidoreductase activity lyase activity heme binding glutathione binding prostaglandin-E synthase activity
          Process(es)
          prostaglandin biosynthetic process cell redox homeostasis positive regulation of transcription, DNA-templated secretion oxidation-reduction process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0