Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
In direct ELISAs, approximately 15% cross-reactivity with recombinant human Marapsin is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Pancreasin is a pancreatic tryptic serine peptidase that cleaves peptides after an arginine residue., Sep 2003].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAP-2; channel-activating protease 2-like protein; EC 3.4.21; EC 3.4.21.4; EC 3.4.21.43; Marapsin; MPNMarapsin; Pancreasin; Serine protease 27
Gene Aliases: CAPH2; marapsin; Mpn; Prss27
UniProt ID: (Mouse) Q8BJR6
Entrez Gene ID: (Mouse) 213171
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support