Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
PRKACG (PRKAC gamma) is a third isoform of the catalytic subunit of cAMP-dependent protein kinase. It was isolated from a human testis cDNA library and was clearly derived from a gene distinct from C-alpha and C-beta and showed tissue-specific expression. Whereas at the amino acid level C-alpha and C-beta showed 93% homology, C-gamma showed only about 80% homology to both C-alpha and C-beta. The PRKACG gene is intronless, contains remnants of a poly(A) tail, is flanked by direct repeats, and is co-linear with the PRKACA gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cAMP-dependent protein kinase catalytic subunit gamma; PKA C-gamma; protein kinase, cAMP-dependent, catalytic, gamma; protein kinase, cAMP-dependent, gamma catalytic subunit; serine(threonine) protein kinase
Gene Aliases: BDPLT19; KAPG; PKACg; PRKACG
UniProt ID: (Human) P22612
Entrez Gene ID: (Human) 5568
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support