Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LPGARYAQLP PAPGLPLERC ALPPLDPGGL KAYPGGECSH LPAVMPAFTV YNGELLYGSP ATTAYYPLKL
Antigen sequence identity: Mouse 77%; Rat 78%
PRDM13 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PR domain containing 13; PR domain zinc finger protein 13; PR domain-containing protein 13; PR-domain containing protein 13; PR-domain zinc finger protein 13
Gene Aliases: MU-MB-20.220; PFM10; PRDM13
UniProt ID: (Human) Q9H4Q3
Entrez Gene ID: (Human) 59336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support