Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MASHVDLLTE LQLLEKVPTL ERLRAAQKRR AQQLKKWAQY EQDLQHRKRK HERKRSTGGR RKKVSFEASV ALLEASLRND AEEVRYFLKN KVSPDLCNED GLTALHQCCI DNFEEIVKLL LSHGANVNAK DNELWTPLHA AATCGHINLV KILVQYGADL LAVNSDGNMP YDLCEDEPTL DVIETCMAYQ GITQEKINEM RVAPEQQMIA DIHCMIAAGQ DLDWIDAQGA TLLHIAGANG YLRAAELLLD HGVRVDVKDW DGWEPLHAAA FWGQMQMAEL LVSHGASLSA RTSMDEMPID LCEEEEFKVL LLELKHKHDV IMKSQLRHKS SLSRRTSSAG SRGKVVRRAS LSDRTNLYRK EYEGEAILWQ RSAAEDQRTS TYNGDIRETR TDQENKDPNP RLEKPVLLSE FPTKIPRGEL DMPVENGLRA PVSAYQYALA NGDVWKVHEV PDYSMAYGNP GVADATPPWS SYKEQSPQTL LELKRQRAAA KLLSHPFLST HLGSSMARTG ESSSEGKAPL IGGRTSPYSS NGTSVYYTVT SGDPPLLKFK APIEEMEEKV HGCCRIS
The protein encoded by this gene is membrane-associated and contains five ankyrin repeats, a protein phosphatase-1-interacting domain, and a carboxy-terminal CAAX box domain. Synthesis of the encoded protein is inhibited by transforming growth factor beta-1. The protein may bind to the membrane through its CAAX box domain and may act as a signaling molecule through interaction with protein phosphatase-1. Alternatively spliced transcript variants encoding different isoforms have been identified in this gene.[provided by RefSeq, Feb 2010].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ankyrin repeat domain protein 4; Ankyrin repeat domain-containing protein 4; CAAX box protein TIMAP; hTIMAP; Protein phosphatase 1 regulatory inhibitor subunit 16B; protein phosphatase 1, regulatory (inhibitor) subunit 16B; protein phosphatase 1, regulatory subunit 16B; TGF-beta-inhibited membrane-associated protein
Gene Aliases: ANKRD4; KIAA0823; PPP1R16B; TIMAP
UniProt ID: (Human) Q96T49
Entrez Gene ID: (Human) 26051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support