Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is specific to PPM1L.
Immunogen sequence: MPAFSTTAA EYVKSRLPEA LKQHLQDYEK DKENSVLSYQ TILEQQILSI DREMLEKLTV SYDEAGTTCL IALLSDKDLT VANVGDSRGV LCDKDGNAIP LSHDHKPYQL KERKRIKRAG GFISFNGSWR VQGILAMSRS LGDYPLKNLN VVIPDPDILT FDLDKLQPEF MILASDGLWD AFSNEEAVRF IKERLDEPHF GAKSIVLQSF YRGCPDNITV MVVKFRNSSK TEEQ (1-233 aa encoded by BC104885)
PPM1L, or PP2CE, belongs to the PP2C group of serine/threonine phosphatases, which are distinguished from other phosphatases by their structure, absolute requirement for Mg(2+) or Mn(2+), and insensitivity to okadaic acid. PP2Cs regulate stress-activated protein kinase (SAPK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PP2C epsilon; PP2C-epsilon; Protein phosphatase 1-like; Protein phosphatase 1L; protein phosphatase 2a, catalytic subunit, epsilon isoform; protein phosphatase 2C epsilon; Protein phosphatase 2C epsilon isoform; Protein phosphatase 2C isoform epsilon; protein phosphatase, Mg2+/Mn2+ dependent, 1L
Gene Aliases: 3222401G13; 5930404J21Rik; AI481720; AW045850; Kiaa4175; mKIAA4175; PP2C; PP2C-epsilon; PP2CE; PP2Cepsilon; PPM1-LIKE; PPM1L; Ppp2ce
UniProt ID: (Human) Q5SGD2, (Mouse) Q8BHN0
Entrez Gene ID: (Human) 151742, (Rat) 310506, (Mouse) 242083
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support