Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MGAFLDKPKT EKHNAHGAGN GLRYGLSSMQ GWRVEMEDAH TAVVGIPHGL EDWSFFAVYD GHAGSRVANY CSTHLLEHIT TNEDFRAAGK SGSALELSVE NVKNGIRTGF LKIDEYMRNF SDLRNGMDRS GSTAVGVMIS PKHIYFINCG DSRAVLYRNG QVCFSTQDHK PCNPREKERI QNAGGSVMIQ RVNGSLAVSR ALGDYDYKCV DGKGPTEQLV SPEPEVYEIL RAEEDEFIIL ACDGIWDVMS NEELCEYVKS RLEVSDDLEN VCNWVVDTCL HKGSRDNMSI VLVCFSNAPK VSDEAVKKDS ELDKHLESRV EEIMEKSGEE GMPDLAHVMR ILSAENIPNL PPGGGLAGKR NVIEAVYSRL NPHRESDGAS DEAEESGSQG KLVEALRQMR INHRGNYRQL LEEMLTSYRL AKVEGEESPA EPAATATSSN SDAGNPVTMQ ESHTESESGL AELDSSNEDA GTKMSGEKI
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PP2C-beta; Protein phosphatase 1B; protein phosphatase 1B (formerly 2C), magnesium-dependent, beta isoform; Protein phosphatase 2C isoform beta; protein phosphatase 2C-like protein; protein phosphatase, Mg2+/Mn2+ dependent, 1B; Ser/Thr protein phosphatase type 2C beta 2 isoform
Gene Aliases: PP2C-beta; PP2C-beta-X; PP2CB; PP2CBETA; PPC2BETAX; PPM1B
UniProt ID: (Human) O75688
Entrez Gene ID: (Human) 5495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support