Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MATAASNPYS ILSSTSLVHA DSAGMQQGSP FRNPQKLLQS DYLQGVPSNG HPLGHHWVTS LSDGGPWSST LATSPLDQQD VKPGREDLQL GAIIHHRSPH VAHHSPHTNH PNAWGASPAP NPSITSSGQP LNVYSQPGFT VSGMLEHGGL TPPPAAASAQ SLHPVLREPP DHGELGSHHC QDHSDEETPT SDELEQFAKQ FKQRRIKLGF TQADVGLALG TLYGNVFSQT TICRFEGLQL SFKNMCKLKP LLNKWLEEAD SSTGSPTSID KIAAQGRKRK KRTSIEVSVK GVLETHFLKC PKPAAQEISS LADSLQLEKE VVRVWFCNRR QKEKRMTPPG DQQPHEVYSH TVKTDTSCHD L
POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-4; Brain-specific homeobox/POU domain protein 4; Oct-9; Octamer-binding protein 9; Octamer-binding transcription factor 9; OTF-9; POU domain, class 3, transcription factor 4
Gene Aliases: BRAIN-4; BRN-4; BRN4; DFN3; DFNX2; OCT-9; OTF-9; OTF9; POU3F4
UniProt ID: (Human) P49335
Entrez Gene ID: (Human) 5456
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support