Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SILSSTSLV HADSAGMQQG SPFRNPQKLL QSDYLQGVPS NGHPLGHHWV TSLSDGGPWS STLATSPLDQ QDVKPGREDL QLGAIIHHRS PHVAHHSPHT NHPNAWGASP APNPSITSSG QPLNVYSQPG FTVSGMLEHG GLTPPPAAAS AQSLHPVLRE PPDHGELGSH HCQ (10-181 aa encoded by BC146551)
POU3F4 (brain-specific homeobox/POU domain protein 4, brain-4, Brn-4, transcription factor 4) is a transcription factor with a POU domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-4; brain-4 class III POU-domain protein; Brain-specific homeobox/POU domain protein 4; Oct-9; Octamer-binding protein 9; Octamer-binding transcription factor 9; OTF-9; POU domain, class 3, transcription factor 4; sex linked fidget
Gene Aliases: BRAIN-4; BRN-4; BRN4; DFN3; DFNX2; OCT-9; OTF-9; OTF9; POU3F4; Slf
UniProt ID: (Human) P49335, (Mouse) P62515
Entrez Gene ID: (Human) 5456, (Mouse) 18994
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support