Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MDSTKEKCDS YKDDLLLRMG LNDNKAGMEG LDKEKINKII MEATKGSRFY GNELKKEKQV NQRIENMMQQ KAQITSQQLR KAQLQVDRFA MELEQSRNLS NTIVHIDMDA FYAAVEMRDN PELKDKPIAV GSMSMLSTSN YHARRFGVRA AMPGFIAKRL CPQLIIVPPN FDKYRAVSKE VKEILADYDP NFMAMSLDEA YLNITKHLEE RQNWPEDKRR YFIKMGSSVE NDNPGKEVNK LSEHERSISP LLFEESPSDV QPPGDPFQVN FEEQNNPQIL QNSVVFGTSA QEVVKEIRFR IEQKTTLTAS AGIAPNTMLA KVCSDKNKPN GQYQILPNRQ AVMDFIKDLP IRKVSGIGKV TEKMLKALGI ITCTELYQQR ALLSLLFSET SWHYFLHISL GLGSTHLTRD GERKSMSVER TFSEINKAEE QYSLCQELCS ELAQDLQKER LKVLYFDMVS LVFKFFNSKM LP
External and internal DNA-damaging agents continually threaten the integrity of genetic material in cells. Although a variety of repair mechanisms exist to remove the resulting lesions, some lesions escape repair and block the replication machinery. Members of the Y family of DNA polymerases, such as POLK, permit the continuity of the replication fork by allowing replication through such DNA lesions. Each Y family polymerase has a unique DNA-damage bypass and fidelity profile. POLK is specialized for the extension step of lesion bypass.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DINB protein; DINP; DNA polymerase kappa; polymerase (DNA directed) kappa
Gene Aliases: DINB1; DINP; POLK; POLQ
UniProt ID: (Human) Q5Q9G8
Entrez Gene ID: (Human) 51426
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support