Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: TNFDELHCKG LTKRNLKRLH FDSPPHLLDI YPRTPHPHEE KVETVEINPK LEVIDLQIST PPVMDILSFD STTTTETSGR TLPDKGEEAC GYFLNPSLMS PECLPTTTKK KTEDMSDLPC ESQRSIPLAV TDALEHIMEQ LNVLTQTVSI LEQRLTLTED KLKDCLENQQ KLFSAVQQKS
Required for ciliogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pix1; POC1 centriolar protein homolog B; Proteome of centriole protein 1B; WD repeat-containing protein 51B
Gene Aliases: CORD20; PIX1; POC1B; TUWD12; WDR51B
UniProt ID: (Human) Q8TC44
Entrez Gene ID: (Human) 282809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support