Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: SREEARAQEA AEFEEMAAWA LAAGRKVKKE PGLAAEVGSA LKAETPNNWN ATEDQHEPTK PLVRRAGAKS RSRRKKQKKN SRQEAVPWKK PKGINSNSTA NLEDPEVGDA ESMAISEPIK GSRKPCVNKE ELALKKPMAK CAWKGPREPP QDARAEAESP GGASESDQDG GHESPPKKKA VAWVSAKNPA PMRKKKKVSL GPVSYVLVDS EDGRKKPVMP KKGPGSRREA SDQKAPRGQQ PAEATASTSR GPKAKPEGSP RRATNESRKV
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Paraneoplastic antigen-like protein 8A; paraneoplastic Ma antigen family-like 1; PNMA-like protein 1
Gene Aliases: PNMA8A; PNMAL1
UniProt ID: (Human) Q86V59
Entrez Gene ID: (Human) 55228
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support