Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MERAESSSTE PAKAIKPIDR KSVHQICSGQ VVLSLSTAVK ELVENSLDAG ATNIDLKLKD YGVDLIEVSD NGCGVEEENF EGLTLKHHTS KIQEFADLTQ VETFGFRGEA LSSLCALSDV TISTCHASAK VGTRLMFDHN GKIIQKTPYP RPRGTTVSVQ QLFSTLPVRH KEFQRNIKKE YAKMVQVLHA YCIISAGIRV SCTNQLGQGK RQPVVCTGGS PSIKENIGSV FGQKQLQSLI PFVQLPPSDS VCEEYGLSCS DALHNLFYIS GFISQCTHGV GRSSTDRQFF FINRRPCDPA KVCRLVNEVY HMYNRHQYPF VVLNISVDSE CVDINVTPDK RQILLQEEKL LLAVLKTSLI GMFDSDVNKL NVSQQPLLDV EGNLIKMHAA DLEKPMVEKQ DQSPSLRTGE EKKDVSISRL REAFSLRHTT ENKPHSPKTP EPRRSPLGQK RGMLSSSTSG AISDKGVLRP QKEAVSSSHG PSDPTDRAEV EKDSGHGSTS VDSEGFSIPD TGSHCSSEYA ASSPGDRGSQ EHVDSQEKAP ETDDSFSDVD CHSNQEDTGC KFRVLPQPTN LATPNTKRFK KEEILSSSDI CQKLVNTQDM SASQVDVAVK INKKVVPLDF SMSSLAKRIK QLHHEAQQSE GEQNYRKFRA KICPGENQAA EDELRKEISK TMFAEMEIIG QFNLGFIITK LNEDIFIVDQ HATDEKYNFE MLQQHTVLQG QRLIAPQTLN LTAVNEAVLI ENLEIFRKNG FDFVIDENAP VTERAKLISL PTSKNWTFGP QDVDELIFML SDSPGVMCRP SRVKQMFASR ACRKSVMIGT ALNTSEMKKL ITHMGEMDHP WNCPHGRPTM RHIANLGVIS QN
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein PMS2; H_DJ0042M02.9; Mismatch repair endonuclease PMS2; PMS1 homolog 2, mismatch repair protein; PMS1 protein homolog 2; PMS2 postmeiotic segregation increased 2; postmeiotic segregation increased 2 nirs variant 6
Gene Aliases: HNPCC4; MLH4; PMS2; PMS2CL; PMSL2
UniProt ID: (Human) P54278
Entrez Gene ID: (Human) 5395
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support