Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: PYNYTSGAAT GWPSMARIAQ STEVLFQGQA SLDCGHGHPD GRRLLLSSSL VEALDVWAGV FSAAAGEGQE RRSPTTTALC LFRMSEIQAR AKRVSWDFK
Receptor for SEMA7A, for smallpox semaphorin A39R, vaccinia virus semaphorin A39R and for herpesvirus Sema protein. Binding of semaphorins triggers cellular responses leading to the rearrangement of the cytoskeleton and to secretion of IL6 and IL8. {ECO:0000250, ECO:0000269; PubMed:20727575}.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD232; CD232 antigen; plexin (semaphorin receptor); Plexin-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; VESPRplexin-C1; Virus-encoded semaphorin protein receptor
Gene Aliases: CD232; PLXN-C1; PLXNC1; VESPR
UniProt ID: (Human) O60486
Entrez Gene ID: (Human) 10154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support