In IHC applications, antigen retrieval using Proteinase K treatment is recommended.
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey, Marmoset (94%) Dog, Panda, Horse (89%) Rat, Elephant (83%).
The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.