Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PIP4K2A Antibodies

          Abnova

          PIP4K2A Monoclonal Antibody (3H5)

          View all (17) PIP4K2A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PIP4K2A Monoclonal Antibody (3H5)

          • Antibody Testing Data (1)
          PIP4K2A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PIP4K2A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PIP4K2A Antibody (H00005305-M08) in WB

          Western Blot analysis of PIP4K2A expression in transfected 293T cell line by PIP4K2A monoclonal antibody (M08), clone 3H5. Lane 1: PIP4K2A transfected lysate(46.2 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PIP4K2A Antibody in Western Blot (WB)

          Product Details

          H00005305-M08

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3H5

          Immunogen

          PIP4K2A (AAH18034, 1 a.a. approximately 406 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.8 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          Immunogen sequence: MATPGNLGSS VLASKTKTKK KHFVAQKVKL FRASDPLLSV LMWGVNHSIN ELSHVQIPVM LMPDDFKAYS KIKVDNHLFN KENMPSHFKF KEYCPMVFRN LRERFGIDDQ DFQNSLTRSA PLPNDSQARS GARFHTSYDK RYIIKTITSE DVAEMHNILK KYHQYIVECH GITLLPQFLG MYRLNVDGVE IYVIVTRNVF SHRLSVYRKY DLKGSTVARE ASDKEKAKEL PTLKDNDFIN EGQKIYIDDN NKKVFLEKLK KDVEFLAQLK LMDYSLLVGI HDVERAEQEE VECEENDGEE EGESDGTHPV GTPPDSPGNT LNSSPPLAPG EFDPNIDVYG IKCHENSPRK EVYFMAIIDI LTHYDAKKKA AHAAKTVKHG AGAEISTVNP EQYSKRFLDF IGHILT

          Target Information

          PIP4K2A lipid kinase or phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, is a member of the phosphatidylinositol-5-phosphate 4-kinase family that catalyzes the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-4,5-bisphosphate. PIP4K2A plays as essential role in the phosphoinositide signal transduction cascades as the precursor to second messengers and is involved in the regulation of secretion, cell proliferation, differentiation, and motility. PIP4K2A may be one of the factors related to the regulation of the beta-globin gene expression and the different levels of Hb H in alpha-thalassemic patients. Diseases associated with PIP4K2A include B-Lymphoblastic Leukemia/Lymphoma With Hyperdiploidy and Bipolar Disorder.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha; 1-phosphatidylinositol-4-phosphate kinase; 1-phosphatidylinositol-4-phosphate-5-kinase; 1-phosphatidylinositol-5-phosphate 4-kinase 2-alpha; Diphosphoinositide kinase 2-alpha; Phosphatidylinositol 5-Phosphate 4-Kinase; Phosphatidylinositol 5-phosphate 4-kinase type II alpha; Phosphatidylinositol 5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-4-phosphate 5-kinase, type II, alpha; phosphatidylinositol-5-phosphate 4-kinase type-2 alpha; phosphatidylinositol-5-phosphate 4-kinase, type II, alpha; PI(5)P 4-kinase type II alpha; PI5P4Kalpha; PIP4KII-alpha; PIP5KIIalpha; PIP5KIII; PtdIns(4)P-5-kinase B isoform; PtdIns(4)P-5-kinase C isoform; PtdIns(5)P-4-kinase isoform 2-alpha; type II phosphatidylinositol-4-phosphate 5-kinase 53 K isoform

          View more View less

          Gene Aliases: PI5P4KA; PIP4K2A; PIP5K2; PIP5K2A; PIP5KII-alpha; PIP5KIIA; PIPK

          View more View less

          UniProt ID: (Human) P48426

          View more View less

          Entrez Gene ID: (Human) 5305

          View more View less

          Function(s)
          ATP binding 1-phosphatidylinositol-4-phosphate 5-kinase activity 1-phosphatidylinositol-5-phosphate 4-kinase activity 1-phosphatidylinositol-3-phosphate 4-kinase activity
          Process(es)
          phosphatidylinositol biosynthetic process regulation of autophagy regulation of phosphatidylinositol 3-kinase signaling megakaryocyte development phosphatidylinositol phosphorylation positive regulation of autophagosome assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-588f58b695-w7k2p:80/100.66.134.75:80.
          git-commit: b8205085541c8e5054bc4cc0f27ee9f88e66ec64
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0