Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Its interaction with PHYH suggests a role in the development of the central system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DYRK1A interacting protein 3; lupus nephritis-associated peptide 1 interacting protein; PAHX-AP1; PAHXAP1; phytanoyl-CoA alpha-hydroxylase associated protein; phytanoyl-CoA hydroxylase interacting protein; Phytanoyl-CoA hydroxylase-associated protein 1; Phytanoyl-CoA hydroxylase-interacting protein
Gene Aliases: AW049870; C630010D02Rik; DYRK1AP3; KIAA0273; Lnap1ip; PAHX-AP; PAHX-AP#1; PAHX-AP1; PAHXAP1; PHYHIP
UniProt ID: (Human) Q92561, (Mouse) Q8K0S0, (Rat) Q568Z9
Entrez Gene ID: (Human) 9796, (Mouse) 105653, (Rat) 290356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support