Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
PCSK7 is likely to represent a ubiquitous endoprotease activity within constitutive secretory pathways and capable of cleavage at the RXXX[KR]R consensus motif.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPC8; Lymphoma proprotein convertase; PC7; PC8; Prohormone convertase 7; prohormone convertase PC7; Proprotein convertase 7; Proprotein convertase 8; proprotein convertase PC7; Proprotein convertase subtilisin/kexin type 7; proprotein convertase subtilisin/kexin type 7 precursor variant 2; Subtilisin/kexin-like protease PC7
Gene Aliases: LPC; PC7; PC8; PCSK7; SPC7
UniProt ID: (Human) Q16549
Entrez Gene ID: (Human) 9159
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support