Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MESKVSEGGL NVTLTIRLLM HGKEVGSIIG KKGETVKKMR EESGARINIS EGNCPERIVT ITGPTDAIFK AFAMIAYKFE EDIINSMSNS PATSKPPVTL RLVVPASQCG SLIGKGGSKI KEIRESTGAQ VQVAGDMLPN STERAVTISG TPDAIIQCVK QICVVMLEAY TIQGQYAIPH PDLTKLHQLA MQQTPFPPLG QTNPAFPGEK LPLHSSEEAQ NLMGQSSGLD ASPPASTHEL TIPNDLIGCI IGRQGTKINE IRQMSGAQIK IANATEGSSE RQITITGTPA NISLAQYLIN ARLTSEVTGM GTL
This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-CP3; PCBP3-overlapping transcript; PCBP3-overlapping transcript 1; Poly(rC)-binding protein 3
Gene Aliases: ALPHA-CP3; PCBP3; PCBP3-OT1; PCBP3OT
UniProt ID: (Human) P57721
Entrez Gene ID: (Human) 54039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support