PA5-27241 targets PAX3 in IF, IHC (P), and WB applications and shows reactivity with Human samples.
The PA5-27241 immunogen is synthetic peptide corresponding to a region within amino acids 442 and 455 of Human PAX3.
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.