FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Opicinumab is under investigation in clinical trial NCT02641041 (Safety, Tolerability, and Pharmacokinetics of Single and Multiple Doses of BIIB033 in Healthy Japanese Participants).