Keep as concentrated solution.
Predicted reactivity: Mouse (92%), Rat (92%), Dog (94%), Rhesus Monkey (100%), Bovine (89%).
Positive Control: 293T, HeLa, HepG2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NCSTN (Nicastrin) is a gene that encodes a type I transmembrane glycoprotein that is an integral component of the multimeric gamma-secretase complex. The encoded protein cleaves integral membrane proteins, including Notch receptors and beta-amyloid precursor protein, and may be a stabilizing cofactor required for gamma-secretase complex assembly. The cleavage of beta-amyloid precursor protein yields amyloid beta peptide, the main component of the neuritic plaque and the hallmark lesion in the brains of patients with Alzheimer's disease; however, the nature of the encoded protein's role in Alzheimer's disease is not known for certain. Mutations in this gene are associated with familial acne inversa. A pseudogene of this gene is present on chromosome 21.