PA1-74133 detects Neutrophil Elastase from human samples.
PA1-74133 has been successfully used in ELISA, immunoelectrophoresis applications.
The PA1-74133 immunogen is Nuetrophil elastase (active-site blocked) from human sputum.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.