Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NALP8 Antibodies

          Abnova

          NLRP8 Polyclonal Antibody, MaxPab™

          View all (3) NALP8 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NLRP8 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          NLRP8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NLRP8 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NLRP8 Antibody (H00126205-B01P) in WB

          Western Blot analysis of NLRP8 expression in transfected 293T cell line by NLRP8 MaxPab polyclonal antibody. Lane 1: NLRP8 transfected lysate(115.28 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NLRP8 Antibody in Western Blot (WB)

          Product Details

          H00126205-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NLRP8 (AAI53175.1, 1 a.a. approximately 1048 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSDVNPPSDT PIPFSSSSTH SSHIPPWTFS CYPGSPCENG VMLYMRNVSH EELQRFKQLL LTELSTGTMP ITWDQVETAS WAEVVHLLIE RFPGRRAWDV TSNIFAIMNC DKMCVVVRRE INAILPTLEP EDLNVGETQV NLEEGESGKI RRYKSNVMEK FFPIWDITTW PGNQRDFFYQ GVHRHEEYLP CLLLPKRPQG RQPKTVAIQG APGIGKTILA KKVMFEWARN KFYAHKRWCA FYFHCQEVNQ TTDQSFSELI EQKWPGSQDL VSKIMSKPDQ LLLLLDGFEE LTSTLIDRLE DLSEDWRQKL PGSVLLSSLL SKTMLPEATL LIMIRFTSWQ TCKPLLKCPS LVTLPGFNTM EKIKYFQMYF GHTEEGDQVL SFAMENTILF SMCRVPVVCW MVCSGLKQQM ERGNNLTQSC PNATSVFVRY ISSLFPTRAE NFSRKIHQAQ LEGLCHLAAD SMWHRKWVLG KEDLEEAKLD QTGVTAFLGM SILRRIAGEE DHYVFTLVTF QEFFAALFYV LCFPQRLKNF HVLSHVNIQR LIASPRGSKS YLSHMGLFLF GFLNEACASA VEQSFQCKVS FGNKRKLLKV IPLLHKCDPP SPGSGVPQLF YCLHEIREEA FVSQALNDYH KVVLRIGNNK EVQVSAFCLK RCQYLHEVEL TVTLNFMNVW KLSSSSHPGS EAPESNGLHR WWQDLCSVFA TNDKLEVLTM TNSVLGPPFL KALAAALRHP QCKLQKLLLR RVNSTMLNQD LIGVLTGNQH LRYLEIQHVE VESKAVKLLC RVLRSPRCRL QCLRLEDCLA TPRIWTDLGN NLQGNGHLKT LILRKNSLEN CGAYYLSVAQ LERLSIENCN LTQLTCESLA SCLRQSKMLT HLSLAENALK DEGAKHIWNA LPHLRCPLQR LVLRKCDLTF NCCQDMISAL CKNKTLKSLD LSFNSLKDDG VILLCEALKN PDCTLQILEL ENCLFTSICC QAMASMLRKN QHLRHLDLSK NAIGVYGILT LCEAFSSQKK REEVIFCIPA WTRITSFSPT PHPPDFTGKS DCLSQINP

          Target Information

          NALP proteins are cytoplasmic proteins that form a subfamily within the larger CATERPILLER family and are thought to play a crucial role in cell proliferation and reproduction. Like all other NALP family members, NALP8 has a C-terminal leucine-rich repeat (LRR) region, an N-terminal Pyrin domain (PYD) followed by a NACHT domain, and a NACHT-associated domain. It maps to chromosome 19q13.42 and is thought to be expressed in testis and ovary, suggesting that NALP8 may be involved in reproduction. Surprisingly, NALP8 seems to be universal in mammals except for rodents, suggesting that the gene may have been lost during evolution.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: NACHT, leucine rich repeat and PYD containing 8; NACHT, LRR and PYD containing protein 8; NACHT, LRR and PYD domains-containing protein 8; NLR family, pyrin domain containing 8; Nucleotide-binding oligomerization domain protein 16; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 8; PYRIN and NACHT-containing protein 4

          View more View less

          Gene Aliases: CLR19.2; NALP8; NLRP8; NOD16; PAN4

          View more View less

          UniProt ID: (Human) Q86W28

          View more View less

          Entrez Gene ID: (Human) 126205

          View more View less

          Function(s)
          molecular_function ATP binding
          Process(es)
          neuron death
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0