PA5-28724 targets NEDL2 in IHC (P) applications and shows reactivity with Human and mouse samples.
The PA5-28724 immunogen is synthetic peptide corresponding to a region within amino acids 1 and 44 of Human NEDL2.
E3 ubiquitin-protein ligase that mediates ubiquitination of TP73. Acts to stabilize TP73 and enhance activation of transcription by TP73.