FIGURE: 1 / 5
PA1-18308 detects Myeloperoxidase from human samples.
PA1-18308 has been successfully used in immunohistochemistry (paraffin and frozen) and Western blot procedures.
The PA1-18308 immunogen is Myeloperoxidase isolated from human polymorphonuclear leucocytes.
Reconstitute with 250 µL of distilled water.
Myeloperoxidase (MPO) is a heme protein that is synthesized during myeloid differentiation and is a major component of neutrophil azurophilic granules. It is produced as a single chain precursor and subsequently cleaved into a light and heavy chain. The mature MPO is a tetramer composed of 2 light chains and 2 heavy chains. This enzyme is responsible for producing hypohalous acids that are central to the microbicidal activity of neutrophils. Diseases that are associated with MPO include Myeloperoxidase Deficiency and Alzheimer's Disease, Familial, 1.