Antibody tested in indirect ELISA
Meloxicam is an oxicam derivative and a non-steroidal anti-inflammatory drug (NSAID) with anti-inflammatory, antipyretic and analgesic activities. Unlike traditional nonselective NSAIDs, meloxicam preferentially inhibits the activity of cyclo-oxygenase II (COX-II), resulting in a decreased conversion of arachidonic acid into prostaglandin precursors. The resulting decrease in prostaglandin synthesis is responsible for the therapeutic effects of meloxicam. [National Center for Biotechnology Information. PubChem Compound Database; CID=54677470].