Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAQGMEPDG YKVVFVRRSP LVLVAVARTR QSAQELAQEL LYIYYQILSL LTGAQLSHIF QQKQNYDLRR LLSGSERITD NLLQLMARDP SFLMGAARCL PLAAAVRDTV SASLQQARAR SLVFSILLAR NQLVALVRRK DQFLHPIDLH LLFNLISSSS SFREGEAWTP VCLPKFNAAG FFHAHISYLE PDTDLCLLLV STDREDFFAV SDCRRRFQER LRKRGAHLAL REALRTPYYS VAQVGIPDLR HFLYKSKSSG LFTSPEIEAP YTSEEEQERL LGLYQYLHSR AHNASRPLKT IYYTGPNENL LAWVTGAFEL YMCYSPLGTK ASAVSAIHKL MRWIRKEEDR LFILTPLTY (1-359 aa encoded by BC047022 )
MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ97088; MGC13272; MON1 homolog A; MON1 secretory trafficking family member A; Vacuolar fusion protein MON1 homolog A
Gene Aliases: MON1A; SAND1
UniProt ID: (Human) Q86VX9
Entrez Gene ID: (Human) 84315
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support