Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MOCOS Antibodies

          Abnova

          MOCOS Monoclonal Antibody (2E8)

          View all (7) MOCOS antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MOCOS Monoclonal Antibody (2E8)

          • Antibody Testing Data (2)
          MOCOS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MOCOS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MOCOS Antibody (H00055034-M05) in WB

          MOCOS monoclonal antibody (M05), clone 2E8. Western Blot analysis of MOCOS expression in human ovarian cancer. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MOCOS Antibody in Western Blot (WB)
          MOCOS Antibody in ELISA (ELISA)

          Product Details

          H00055034-M05

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          1 ng/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2E8

          Immunogen

          MOCOS (AAH12079.1, 1 a.a. approximately 888 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAGAAAESGR ELWTFAGSRD PSAPRLAYGY GPGSLRELRA REFSRLAGTV YLDHAGATLF SQSQLESFTS DLMENTYGNP HSQNISSKLT HDTVEQVRYR ILAHFHTTAE DYTVIFTAGS TAALKLVAEA FPWVSQGPES SGSRFCYLTD SHTSVVGMRN VTMAINVISI PVRPEDLWSA EERGASASNP DCQLPHLFCY PAQSNFSGVR YPLSWIEEVK SGRLRPVSTP GKWFVLLDAA SYVSTSPLDL SAHQADFVPI SFYKIFGFPT GLGALLVHNR AAPLLRKTYF GGGTASAYLA GEDFYIPRQS VAQRFEDGTI SFLDVIALKH GFDTLERLTG GMENIKQHTF TLAQYTYMAL SSLQYPNGAP VVRIYSDSEF SSPEVQGPII NFNVLDDKGN IIGYSQVDKM ASLYNIHLRT GCFCNTGACQ RHLGISNEMV RKHFQAGHVC GDNMDLIDGQ PTGSVRISFG YMSTLDDVQA FLRFIIDTRL HSSGDWPVPQ AHADTGETGA PSADSQADVI PAVMGRRSLS PQEDALTGSR VWNNSSTVNA VPVAPPVCDV ARTQPTPSEK AAGVLEGALG PHVVTNLYLY PIKSCAAFEV TRWPVGNQGL LYDRSWMVVN HNGVCLSQKQ EPRLCLIQPF IDLRQRIMVI KAKGMEPIEV PLEENSERTQ IRQSRVCADR VSTYDCGEKI SSWLSTFFGR PCHLIKQSSN SQRNAKKKHG KDQLPGTMAT LSLVNEAQYL LINTSSILEL HRQLNTSDEN GKEELFSLKD LSLRFRANII INGKRAFEEE KWDEISIGSL RFQVLGPCHR CQMICIDQQT GQRNQHVFQK LSESRETKVN FGMYLMHASL DLSSPCFLSV GSQVLPVLKE NVEGHDLPAS EKHQDVTS

          Target Information

          MOCOS sulfurates the molybdenum cofactor of xanthine dehydrogenase (XDH), which is required for their enzymatic activities (Ichida et al., 2001).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: MCS; MOCOS; Molybdenum cofactor sulfurase; Molybdenum cofactor sulfurtransferase; MOS

          View more View less

          Gene Aliases: HMCS; MCS; MOCOS; MOS

          View more View less

          UniProt ID: (Human) Q96EN8

          View more View less

          Entrez Gene ID: (Human) 55034

          View more View less

          Function(s)
          protein binding Mo-molybdopterin cofactor sulfurase activity transferase activity lyase activity molybdenum ion binding pyridoxal phosphate binding
          Process(es)
          Mo-molybdopterin cofactor biosynthetic process molybdopterin cofactor biosynthetic process molybdopterin cofactor metabolic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0