Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is specific to MMP26.
Immunogen sequence: ADHKGWDFV EGYFHQFFLT KKESPLLTQE TQTQLLQQFH RNGTDLLDMQ MHALLHQPHC GVPDGSDTSI SPGRCKWNKH TLTYRIINYP HDMKPSAVKD SIYNAVSIWS NVTPLIFQQV QNGDADIKVS FWQWAHEDGW PFDGPGGILG HAFLPNSGNP GVVHFDKNEH WSASDTGYNL FLVATHEIGH SLGLQHSGNQ SSIMYPTYWY HDPRTFQLSA DDIQRIQHLY GEKCSSDIP (24-262 aa encoded by BC101541)
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The encoded protein degrades type IV collagen, fibronectin, fibrinogen, casein, vitronectin, alpha 1-antitrypsin, alpha 2-macroglobulin, and insulin-like growth factor-binding protein 1, and activates MMP9 by cleavage. The protein differs from most MMP family members in that it lacks a conserved C-terminal protein domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endometase; matrilysin 2; Matrilysin-2; matrix metallo protease; matrix metalloproteinase 26; Matrix metalloproteinase-26; MGC126590; MGC126592; MMP; MMP-26; MMPs
Gene Aliases: MMP26
UniProt ID: (Human) Q9NRE1
Entrez Gene ID: (Human) 56547
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support