Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HRX Antibodies

          QED Bioscience

          MLL Monoclonal Antibody (N4.4)

          View all (15) HRX antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MLL Monoclonal Antibody (N4.4)

          • Antibody Testing Data (1)
          MLL Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MLL Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MLL Antibody (34041-100UG) in WB

          Immunoblotting: use at 2 µg/mL. A band of ~300 kDa is detected. K562 cell nuclear extract with #34041 at 2 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MLL Antibody in Western Blot (WB)

          Product Details

          34041-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          -

          Immunoprecipitation (IP)

          2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N4.4

          Immunogen

          Maltose-binding fusion protein containing MLL amino acids 161-356 (Uniprot #Q03164).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.0 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          KMT2A is a histone methyltransferase that plays an essential role in early development and hematopoiesis. It is a catalytic subunit of the MLL1/MLL complex, a multiprotein complex that mediates both methylation of 'Lys-4' of histone H3 (H3K4me) complex and acetylation of 'Lys-16' of histone H4 (H4K16ac). In the MLL1/MLL complex, KMT2A specifically mediates H3K4me, a tag for epigenetic transcriptional activation. KMT2A has weak methyltransferase activity by itself, and requires other components of the MLL1/MLL complex to obtain full methyltransferase activity. It has no activity toward histone H3 phosphorylated on 'Thr-3', less activity toward H3 dimethylated on 'Arg-8' or 'Lys-9', and has higher activity toward H3 acetylated on 'Lys-9'. Required for transcriptional activation of HOXA9 and promotes PPP1R15A-induced apoptosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALL-1; CDK6/MLL fusion protein; CXXC-type zinc finger protein 7; Cysteine methyltransferase KMT2A; Histone-lysine N-methyltransferase 2A; histone-lysine N-methyltransferase MLL; lysine (K)-specific methyltransferase 2A; Lysine N-methyltransferase 2A; mixed lineage leukemia 1; MLL-AF4 der(11) fusion protein; MLL/ENL fusion protein; MLL/GAS7 fusion protein; MLL/GMPS fusion protein; MLL/hCDCrel fusion protein; Myeloid/lymphoid or mixed-lineage leukemia; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); myeloid/lymphoid or mixed-lineage leukemia 1; Myeloid/lymphoid or mixed-lineage leukemia protein 1; rearranged MLL protein; trithorax Drosophila; Trithorax-like protein; Zinc finger protein HRX

          View more View less

          Gene Aliases: 6430520K01; ALL-1; ALL1; CXXC7; HRX; HTRX; HTRX1; KMT2A; mKIAA4050; MLL; MLL-AF9; MLL/GAS7; MLL1; MLL1A; TET1-MLL; TRX1; WDSTS

          View more View less

          UniProt ID: (Human) Q03164

          View more View less

          Entrez Gene ID: (Human) 4297, (Mouse) 214162

          View more View less

          Function(s)
          core promoter sequence-specific DNA binding AT DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding histone-lysine N-methyltransferase activity histone methyltransferase activity (H3-K4 specific) identical protein binding protein homodimerization activity transcription regulatory region DNA binding unmethylated CpG binding lysine-acetylated histone binding DNA binding methyltransferase activity transferase activity metal ion binding
          Process(es)
          DNA methylation transcription, DNA-templated regulation of transcription, DNA-templated protein complex assembly negative regulation of cell proliferation anterior/posterior pattern specification chromatin modification methylation positive regulation of transporter activity circadian regulation of gene expression embryonic hemopoiesis histone H4-K16 acetylation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter rhythmic process histone H3-K4 methylation regulation of histone H3-K4 methylation positive regulation of histone H3-K4 methylation regulation of histone H3-K14 acetylation histone H3-K4 trimethylation regulation of histone H3-K9 acetylation positive regulation of cellular response to drug transcription from RNA polymerase II promoter apoptotic process visual learning post-embryonic development peptidyl-lysine monomethylation exploration behavior response to potassium ion histone H3-K4 dimethylation regulation of short-term neuronal synaptic plasticity spleen development homeostasis of number of cells within a tissue membrane depolarization definitive hemopoiesis regulation of histone H3-K27 acetylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66966984d8-2vwff:80/100.66.134.75:80.
          git-commit: 4f7892df6eac4c3aaa23131f1084e28be0ddeb54
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0