Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
Sequence of this protein is as follows: MKAPAVLAPG ILVLLFTLVQ RSNGECKEAL AKSEMNVNMK YQLPNFTAET PIQNVILHEH HIFLGATNYI YVLNEEDLQK VAEYKTGPVL EHPDCFPCQD CSSKANLSGG VWKDNINMAL VVDTYYDDQL ISCGSVNRGT CQRHVFPHNH TADIQSEVHC IFSPQIEEPS QCPDCVVSAL GAKVLSSVKD RFINFFVGNT INSSYFPDHP LHSISVRRLK ETKDGFMFLT DQSYIDVLPE FRDSYPIKYV HAFESNNFIY FLTVQRETLD AQTFHTRIIR FCSINSGLHS YMEMPLECIL TEKRKKRSTK KEVFNILQAA YVSKPGAQLA RQIGASLNDD ILFGVFAQSK PDSAEPMDRS AMCAFPIKYV NDFFNKIVNK NNVRCLQHFY GPNHEHCFNR TLLRNSSGCE ARRDEYRTEF TTALQRVDLF MGQFSEVLLT SISTFIKGDL TIANLGTSEG RFMQVVVSRS GPSTPHVNFL LDSHPVSPEV IVEHTLNQNG YTLVITGKKI TKIPLNGLGC RHFQSCSQCL SAPPFVQCGW CHDKCVRSEE CLSGTWTQQI CLPAIYKVFP NSAPLEGGTR LTICGWDFGF RRNNKFDLKK TRVLLGNESC TLTLSESTMN TLKCTVGPAM NKHFNMSIII SNGHGTTQYS TFSYVDPVIT SISPKYGPMA GGTLLTLTGN YLNSGNSRHI SIGGKTCTLK SVSNSILECY TPAQTISTEF AVKLKIDLAN RETSIFSYRE DPIVYEIHPT KSFISGGSTI TGVGKNLNSV SVPRMVINVH EAGRNFTVAC QHRSNSEIIC CTTPSLQQLN LQLPLKTKAF FMLDGILSKY FDLIYVHNPV FKPFEKPVMI SMGNENVLEI KGNDIDPEAV KGEVLKVGNK SCENIHLHSE AVLCTVPNDL LKLNSELNIE WKQAISSTVL GKVIVQPDQN FTGLIAGVVS ISTALLLLLG FFLWLKKRKQ IKDLGSELVR YDARVHTPHL DRLVSARSVS PTTEMVSNES VDYRATFPED QFPNSSQNGS CRQVQYPLTD MSPILTSGDS DISSPLLQNT VHIDLSALNP ELVQAVQHVV IGPSSLIVHF NEVIGRGHFG CVYHGTLLDN DGKKIHCAVK SLNRITDIGE VSQFLTEGII MKDFSHPNVL SLLGICLRSE GSPLVVLPYM KHGDLRNFIR NETHNPTVKD LIGFGLQVAK GMKYLASKKF VHRDLAARNC MLDEKFTVKV ADFGLARDMY DKEYYSVHNK TGAKLPVKWM ALESLQTQKF TTKSDVWSFG VLLWELMTRG APPYPDVNTF DITVYLLQGR RLLQPEYCPD PLYEVMLKCW HPKAEMRPSF SELVSRISAI FSTFIGEHYV HVNATYVNVK CVAPYPSLLS SEDNADDEVD TRPASFWETS
MET (cMET) is a receptor-like tyrosine kinase whose dysregulation has been linked to many types of human malignancies. After activation of the ligand, MET interacts with the PI3-kinase subunit PIK3R1, PLCG1, SRC, GRB2, and STAT3. There interactions lead to the activation of signaling cascades including RAS-ERK, PI3, kinase-AKT, and PLCgamma-PKC. MET plays a role in embryonic development including gastrulation, development of muscles and neurons, angiogenesis, and kidney formation. It also plays a role in adults including wound healing, organ regeneration, and tissue remodeling. MET has been linked to cancers including gastric, renal, and breast; therefore, making it a target for cancer therapeutics and diagnostic testing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cMet; EC 2.7.10.1; Hepatocyte growth factor receptor; HGF receptor; HGF-SF receptor; HGF/SF receptor; oncogene MET; Proto-oncogene c-Met; sc MET; Scatter factor receptor; SF receptor; soluble c met; Tyrosine-protein kinase Met
Gene Aliases: AUTS9; c-Met; DFNB97; HGFR; MET; RCCP2
UniProt ID: (Human) P08581
Entrez Gene ID: (Human) 4233
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support